Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Dog IgG F(c) fragment, FITC, Polyclonal, Secondary Antibody, Abnova™

Catalog No. 89137088 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-137-088 1500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-137-088 Supplier Abnova Corporation Supplier No. PAB10545
Add to Cart
Add to Cart

Specifications

Antigen Rabbit anti-Dog IgG F(c) fragment
Applications Flow Cytometry, Immunofluorescence
Classification Polyclonal
Conjugate FITC
Description Rabbit anti-dog IgG recognizes dog IgG F(c) fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Fluorescein.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Host Species Rabbit
Immunogen Dog IgG F(c) fragment
Purification Method This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities.
Quantity 1500 μg
Primary or Secondary Secondary
Target Species Dog
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.