Learn More
Immunology Consultants Affinity Purified Rabbit anti-Dog IgG Fc Antibody, 1.0 mg
SDP
Catalog No. 502248891
Encompass_Preferred
Rabbits were immunized with highly purified dog IgG Fc fragment and the resulting antiserum was collected. Antibody was solid phase adsorbed and immunoaffinity purified off a dog IgG containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with dog IgG and not dog IgG Fab fragmentas determined by IEP and ELISA techniques. It is suitable for blotting, ELISA, and IHC applications. This antibody may cross-react with IgG from other species. Optimal working dilutions should be determined experimentally by the investigator.
This item is not returnable. View return policy
Catalog No. 50-224-8891 Supplier Immunology Consultants Supplier No. RGFC40A
Rabbits were immunized with highly purified dog IgG Fc fragment and the resulting antiserum was collected. Antibody was solid phase adsorbed and immunoaffinity purified off a dog IgG containing immunosorbent. Antibody concentration was determined using an absorbance at 280 nm: 1.4 equals 1.0 mg of IgG. This antibody reacts with dog IgG and not dog IgG Fab fragmentas determined by IEP and ELISA techniques. It is suitable for blotting, ELISA, and IHC applications. This antibody may cross-react with IgG from other species. Optimal working dilutions should be determined experimentally by the investigator.