Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Dog IgM mu heavy chain,Polyclonal Secondary Antibody, Abnova™

Catalog No. 89138883 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-883 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-883 Supplier Abnova Corporation Supplier No. PAB9186
Add to Cart
Add to Cart

Specifications

Antigen Rabbit anti-Dog IgM mu heavy chain
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit anti-dog IgG recognizes dog IgM mu heavy chain. This secondary antibody was purified using a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized
Host Species Rabbit
Immunogen Dog IgM mu heavy chain
Purification Method This product is an IgG fraction antibody purified from monospecific antiserum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography followed by extensive dialysis against the buffer stated above.
Quantity 50 mg
Primary or Secondary Secondary
Test Specificity No reaction was observed against dog IgG.
Target Species Dog
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with 5mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.