Learn More
Rabbit anti-Dog serum proteins, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Rabbit anti-Dog serum proteins |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit anti-dog IgG recognizes dog serum proteins. This secondary antibody was purified using delipidation and defibrination. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized |
| Host Species | Rabbit |
| Immunogen | Dog serum proteins |
| Purification Method | This product was prepared from polyspecific antiserum by a delipidationand defibrination. Assay by immunoelectrophoresis resulted in a multipleprecipitin arcs against dog serum. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.