Learn More
Invitrogen™ Rabbit anti-Donkey IgG (H+L) Secondary Antibody, DyLight™ 800
Shop All Thermo Scientific ProductsDescription
Rabbit anti-Donkey IgG (H+L) Secondary Antibody for Flow, ICC/IF, IHC, IP, Western Blot
Specifications
Specifications
| Antigen | Donkey IgG (H+L) |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | DyLight 800 |
| Formulation | PBS with 0.2% BSA and 0.09% sodium azide; pH 6.8 to 7.4 |
| Host Species | Rabbit |
| Immunogen | Donkey IgG-heavy and light chain. |
| Purification Method | Antigen affinity chromatography |
| Quantity | 500 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.