Learn More
Ibt Bioservices IBT Rabbit anti-Ebola Virus Glycoprotein pAb, Cat # 0301-015, Rabbit, Polyclonal, Ebola Virus, Ebola glycoprotein, Zaire, Affinity purified, ELISA, Western Blot, Liquid, Storage: -80°C
SDP
Supplier:  Ibt Bioservices 0301015
Encompass_Preferred
Affinity purified rabbit polyclonal antibody reactive to EBOV GP. The antibody detects GP in virus-like particles (VLP) and recombinant GP without the transmembrane region (rGPdTM) in Western blot and ELISA.Supplied in PBS in frozen format with no preservatives added. Store at +4°C for 2-3 weeks; -80°C for long-term
Catalog No. NC0963565