Learn More
Rabbit anti-Equine IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alkaline Phosphatase (AP) |
| Dilution | Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:20000, Immunohistochemistry 1:200 - 1:1000 |
| Gene Alias | immunoglobulin heavy constant gamma 1 (G1m marker) |
| Host Species | Rabbit |
| Immunogen | Horse IgG whole molecule |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.5 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.