Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Feline IgG F(ab')2 Secondary Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB484023
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB484023 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB484023 Supplier R&D Systems Supplier No. NBP172719PCP

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG F(ab')2 Polyclonal specifically detects IgG F(ab')2 in Feline samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG F(ab')2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PerCP
Formulation PBS
Host Species Rabbit
Immunogen Cat IgG F(ab')2 fragment
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Feline
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.