Learn More
Rabbit anti-Feline IgM Heavy Chain Secondary Antibody, HRP, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgM |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | HRP |
| Dilution | Western Blot 1:1000 - 1:10000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:500 - 1:2500, Immunohistochemistry-Paraffin 1:500 - 1:2500, Immunohistochemistry-Frozen 1:500 - 1:2500 |
| Formulation | Lyophilized from 0.02M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2, with 10Mg/ml Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Gentamicin Sulfate |
| Gene Alias | AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH |
| Host Species | Rabbit |
| Immunogen | Feline IgM mu heavy chain |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.