Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Fish Immunoglobulin Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP268352
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-68352 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-68352 Supplier R&D Systems Supplier No. NBP268352

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunoglobulin Polyclonal specifically detects Immunoglobulin in Fish samples. It is validated for ELISA, Immunofluorescence, Western Blot.

Specifications

Antigen Immunoglobulin
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, ELISA 1:5000 - 1:10000, Immunofluorescence
Formulation Serum with 0.09% Sodium Azide
Host Species Rabbit
Immunogen Rainbow trout and Atlantic salmon immunoglobulins.
Purification Method Whole Antiserum
Quantity 50 μL
Primary or Secondary Secondary
Test Specificity Rabbit anti Salmonid Ig antibody recognizes salmonid immunoglobulin, binding to Ig of the Salmo, Oncorhynchus and Salvelinus genera
Target Species Fish
Content And Storage Store at 4°C.
Form Antisera
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.