Learn More
ACROBiosystems Rabbit Anti-FITC 10mg
Catalog No. 502534414
Encompass_Preferred
The Anti-FITC Antibody coupled Magnetic Beads are 2.8 µm superparamagnetic particles covalently coupled to a highly affinity Monoclonal anti-FITC antibody. The beads can be used to capture the FITC Antibody in Chemiluminescence procedures. The Anti-FITC Antibody coupled Magnetic Beads is easy to capture the FITC labeled proteins and the bounded proteins have no activity lost this ready to use products could greatly save your protein coupling time and hassle and help us get the best performance and highly reproducible results.
Catalog No. 50-253-4414 Supplier ACROBiosystems Supplier No. MPCA00910MG
The Anti-FITC Antibody coupled Magnetic Beads are 2.8 µm superparamagnetic particles covalently coupled to a highly affinity Monoclonal anti-FITC antibody. The beads can be used to capture the FITC Antibody in Chemiluminescence procedures. The Anti-FITC Antibody coupled Magnetic Beads is easy to capture the FITC labeled proteins and the bounded proteins have no activity lost this ready to use products could greatly save your protein coupling time and hassle and help us get the best performance and highly reproducible results.