Learn More
GenDEPOT Rabbit anti-Goat IgG
Catalog No. 501904211
Encompass_Preferred
Affinity purified antibody isolated from a pool of serum from rabbits immunized with purified rabbit IgG was labeled with peroxidase by the periodate method of Nakane and Kawaoi.
Catalog No. 50-190-4211 Supplier GenDEPOT Supplier No. SA007500
Affinity purified antibody isolated from a pool of serum from rabbits immunized with purified rabbit IgG was labeled with peroxidase by the periodate method of Nakane and Kawaoi.