Learn More
Cayman Chemical Rabbit Anti-Goat IgG Eu-W1024
Supplier: Cayman Chemical 1657450 UG
Concentration: 1 mg/ml Antibody: Rabbit poyclonal anti-Goat IgG Excitation max. λ: 340 nm Emission max. λ: 615 nm Uses: TR-FRET and High throughput screening (HTS) applications Form: Conjugate supplied as a liquid in 50 mn Tris (pH 7.8) + 0.9% NaCl + 0.05% NaN3