Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Goat IgG Fc Secondary Antibody, Allophycocyanin, R&D Systems™
SDP

Catalog No. NB483732
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB483732 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB483732 Supplier R&D Systems Supplier No. NBP174860APC

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Goat samples. It is validated for Chemiluminescence Immunoassay, ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Western Blot.

Specifications

Antigen IgG Fc
Applications Chemiluminescence Immunoassay (CIA), ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunomicroscopy, Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Host Species Rabbit
Immunogen Purified Goat IgG Fc fragment
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.