Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit anti-Goat IgG (H+L) Secondary Antibody, Biotin

Catalog No. PI31732 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31732 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31732 Supplier Invitrogen™ Supplier No. 31732
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Secondary Antibody

Product # 31732 has been successfully used in Western blot, IF, ICC, IHC, ELISA and ISH applications. Product # 31732 reacts with the heavy chains of goat IgG and with the light chains common to most goat immunoglobulins, but does not react against non-immunoglobulin serum proteins. Based on solid-phase enzyme immunoassy, this antibody cross-reacts with sheep IgG (50%) bovine IgG (25%) bovine IgM, goat IgM, sheep IgM (6%) cat IgG, dog IgG (1%) and it has minimal cross-reactivity with immunoglobulins from other species.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat IgG (H+L)
Applications ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Biotin
Formulation PBS with 3mg/mL BSA and 0.08% sodium azide; pH 7.8
Host Species Rabbit
Purification Method Affinity chromatography
Quantity 1.5 mg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.