Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Goat IgG (H+L) Secondary Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB483736
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB483736 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB483736 Supplier R&D Systems Supplier No. NBP174825PE

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Goat samples. It is validated for ELISA, Flow Cytometry, Immunoassay, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG (H+L)
Applications ELISA, Flow Cytometry, Immunoassay, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Host Species Rabbit
Immunogen This Rabbit anti-Goat IgG (H+L) Secondary Antibody was developed against purified goat IgG (H&L).
Purification Method Affinity purified
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Goat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.