Learn More
Immunology Consultants anti-Goat IgG h+l Antibody (Rabbit) - Affinity Purified
SDP
Catalog No. 502248872
Encompass_Preferred
RGHL‑50A is an affinity‑purified rabbit antibody targeting goat IgG heavy and light chains, designed for use as a secondary antibody in immunoassays. This reagent provides strong specificity, low background, and reliable performance in ELISA, Western blotting, and other detection platforms. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑50A is a dependable choice for researchers working with goat primary antibodies.
This item is not returnable. View return policy
Catalog No. 50-224-8872 Supplier Immunology Consultants Supplier No. RGHL50A
RGHL‑50A is an affinity‑purified rabbit antibody targeting goat IgG heavy and light chains, designed for use as a secondary antibody in immunoassays. This reagent provides strong specificity, low background, and reliable performance in ELISA, Western blotting, and other detection platforms. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑50A is a dependable choice for researchers working with goat primary antibodies.