Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Rabbit anti-Goat, HRP, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. AP106PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP106PMI 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP106PMI Supplier MilliporeSigma Supplier No. AP106P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Rabbit anti-Goat IgG Antibody, HRP conjugate is an antibody against Goat IgG for use in ELISA, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit anti-Goat IgG
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate HRP
Formulation Purified rabbit IgG conjugated to horseradish peroxidase in buffer containing 0.01M Sodium Phosphate, 0.15M NaCl, pH 7.1 with 15mg/mL BSA and 0.01% thimerosal. Lyophilized.
Host Species Rabbit
Purification Method Immunoaffinity chromatography using antigen coupled to sepharose beads.
Quantity 1.5 mL
Primary or Secondary Secondary
Target Species Goat
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.