Learn More
Bioworld Rabbit Anti-Goat IgG Antibody (Polyclonal IgG) - Nano-activeGold™, 0.5 mL
Supplier:  Bioworld 201000071
Encompass_Preferred
Rabbit Anti-Goat IgG Antibody (Polyclonal IgG) - Nano-activeGold ™Cat#20100007DetailsHost:RabbitIsotype:IgGClass:PolyclonalImmunogen:Goat IgGConjugate:Nano-activeGold ™Form InformationForm:LiquidConcentration:2 mg/mLPurification:Protein A ChromatrographyStorage Buffer:PBS with 0.1% BSAPreservative:Upon Request (0.02% Sodium Azide)Storage:Shipped at 4°C. Upon delivery aliquot and store at -20°C. Avoid freeze / thaw cycles.Usage RecommendationsWestern Blot (WB):1:3000-1:5000ELISA:1:150-1:400Immunoprecipitation (IP):1:350 - 1:500
Catalog No. 50-197-477