Learn More
Cayman Chemical Rabbit Anti-Goat IgG SureLigh
SDP
Supplier:  Cayman Chemical 165971 MG
Encompass_Preferred
Rabbit anti-Goat conjugated to SureLight Allophycocyanin Antibody: Rabbit polyclonal anti-Goat IgG Dye: SureLight Allophycocyanin (APC) Excitation max. λ: 650 nm Emission max. λ: 660 nm Uses: Flow cytometry and fluorescence-activated cell sorting
Catalog No. 50-296-6537