Learn More
Cayman Chemical Rabbit Anti-Goat IgG SureLigh
SDP
Supplier:  Cayman Chemical 165691 MG
Encompass_Preferred
Rabbit anti-Goat IgG conjugated to SureLight PerCPAntibody: Rabbit polyclonal anti-Goat IgG Dye: Peridinin-Chlorophyll (PerCP) Excitation max. λ: 483 nm Emission max. λ: 673 nm Uses: Flow Cytometry and Protein microarrays Form: Conjugate supplied as a lyophilized powder.
Catalog No. 50-296-6817