Learn More
Cayman Chemical Rabbit Anti-Goat IgG SureLigh
SDP
Catalog No. 502966535
Encompass_Preferred
Rabbit anti-Goat IgG, secondary antibody, conjugated to SureLight R-PE Antibody: Rabbit polyclonal anti-goat IgG Dye: SureLight R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays
Catalog No. 50-296-6535 Supplier Cayman Chemical Supplier No. 166561 MG
Rabbit anti-Goat IgG, secondary antibody, conjugated to SureLight R-PE Antibody: Rabbit polyclonal anti-goat IgG Dye: SureLight R-phycoerythrin Excitation max. λ: 565>498 nm Emission max. λ: 578 nm Uses: Flow cytometry, fluorescence-activated cell sorting, and protein microarrays