Learn More
Rabbit anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG (H+L) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alkaline Phosphatase (AP) |
| Dilution | Western Blot 1:200 - 1:1000, ELISA 1:5500, Immunohistochemistry 1:200 - 1:1000 |
| Host Species | Rabbit |
| Immunogen | Golden Syrian Hamster IgG, whole molecule |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.