Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Golden Syrian Hamster IgG (H+L) Secondary Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB1206779
Change view
Click to view available options
Quantity:
1.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206779 1.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206779 Supplier R&D Systems Supplier No. NB1206779

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG (H+L) Polyclonal specifically detects IgG (H+L) in Golden Syrian Hamster samples. It is validated for Flow Cytometry, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate FITC
Dilution Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000
Host Species Rabbit
Immunogen Hamster IgG whole molecule
Purification Method Immunogen affinity purified
Quantity 1.5 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Test Specificity Prepared from monospecific antiserum by immunoaffinity chromatography using Hamster IgG coupled to agarose. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Rabbit Serum, Hamster IgG and Hamster Serum.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Golden Syrian Hamster
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.