Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Guinea Pig IgG Fc Secondary Antibody, Novus Biologicals™
SDP

Catalog No. NBP172735
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP172735 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP172735 Supplier R&D Systems Supplier No. NBP172735

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG Fc Polyclonal specifically detects IgG Fc in Guinea Pig samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG Fc
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.69 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000
Host Species Rabbit
Immunogen Guinea Pig IgG F(c) fragment
Purification Method Affinity Purified
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity No reaction was observed against Guinea Pig IgG F(ab) .
Target Species Guinea Pig
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.