Learn More
Rabbit anti-Guinea Pig IgG (H+L) Secondary Antibody, Rhodamine, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG |
| Applications | Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Rhodamine |
| Dilution | Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000-1:5000, Fluorophore-linked immunosorbent assay 1:10000-1:50000 |
| Gene Alias | immunoglobulin heavy constant gamma 1 (G1m marker) |
| Host Species | Rabbit |
| Immunogen | Guinea Pig IgG whole molecule |
| Purification Method | Multi-step |
| Quantity | 1 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.