Learn More
Abcam Key Features:Rabbit Anti-Horse IgG H&LHost species: RabbitSuitable for: ELISA, WB, IHC-P, IHC-Fr, IP, Immunodiffusion, Conjugation, ICC/IF.ABCAM LTD mfg# AB6700
Supplier:  Abcam AB67001MG
Encompass_Preferred
Key Features:Rabbit Anti-Horse IgG H&LHost species: RabbitSuitable for: ELISA, WB, IHC-P, IHC-Fr, IP, Immunodiffusion, Conjugation, ICC/IF.ABCAM LTD mfg# AB6700
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2133865