Learn More
Immunology Consultants anti-Horse Whole Serum Antibody (Rabbit) - Antiserum
SDP
Catalog No. 502248967
Encompass_Preferred
This rabbit antiserum against horse whole serum provides broad, multi‑epitope recognition of equine serum proteins. Ideal for ELISA, immunodiffusion, and veterinary diagnostic development, it delivers consistent performance across diverse assay formats. Produced by Immunology Consultants, this 1 ml reagent supports robust detection of horse serum components in both purified and complex samples
This item is not returnable. View return policy
Catalog No. 50-224-8967 Supplier Immunology Consultants Supplier No. RWS70
This rabbit antiserum against horse whole serum provides broad, multi‑epitope recognition of equine serum proteins. Ideal for ELISA, immunodiffusion, and veterinary diagnostic development, it delivers consistent performance across diverse assay formats. Produced by Immunology Consultants, this 1 ml reagent supports robust detection of horse serum components in both purified and complex samples