Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Human Ig kappa light chain, Clone: RM126, Secondary Antibody,

Catalog No. 041660MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
04-166-0MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-166-0MI Supplier MilliporeSigma™ Supplier No. 041660
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

Specifically detects Ig kappa light chain Clone: RM126 in Human samples, and it is validated for ELISA, Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig kappa light chain
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone RM126
Conjugate Unconjugated
Formulation Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Host Species Rabbit
Immunogen Human IgG
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary, Control Antibodies
Primary or Secondary Secondary
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.