Learn More
Rabbit anti-Human IgA Heavy Chain Secondary Antibody, DyLight 350, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgA Heavy Chain |
| Applications | Dot Blot, ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Formulation | 50 mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | Human IgA alpha heavy chain |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.