Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Human IgA Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NBP173613
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP173613 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP173613 Supplier R&D Systems Supplier No. NBP173613

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgA Heavy Chain Polyclonal specifically detects IgA Heavy Chain in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.

Specifications

Antigen IgA Heavy Chain
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate HRP
Dilution Western Blot 1:1000 - 1:5000, ELISA 1:56000, Immunohistochemistry 1:500 - 1:2500
Gene Alias Immunoglobulin A
Gene Symbols IGHA1
Host Species Rabbit
Immunogen Human IgA a alpha chain
Purification Method Affinity Purified
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3493
Test Specificity Specificity was confirmed by ELISA minimal cross reactivity against other Human heavy or light chain isotypes.
Reconstitution Reconstitute with 1.0 ml deionized water (or equivalent).
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgA
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.