Learn More
Rabbit anti-Human IgG Fc Secondary Antibody, Texas Red, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG Fc |
| Applications | Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Texas Red |
| Dilution | Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 |
| Host Species | Rabbit |
| Immunogen | Rabbit anti-Human IgG Fc fragment Secondary Antibody was produced by repeated immunization with Human IgG F(c) fragment in rabbit. |
| Purification Method | Affinity Purified |
| Quantity | 2 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.