Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Human IgG heavy chain, Clone: RM116, Secondary Antibody,

Catalog No. 041650MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
04-165-0MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-165-0MI Supplier MilliporeSigma™ Supplier No. 041650
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifically detects IgG heavy chain Clone: RM116 in Human samples, and it is validated for ELISA, Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG heavy chain
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone RM116
Conjugate Unconjugated
Formulation Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Host Species Rabbit
Immunogen Human IgG
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary, Control Antibodies
Primary or Secondary Secondary
Test Specificity This antibody reacts to the Fc region of all gamma heavy chains of human immunoglobulins, including γ1, γ2, γ3, and γ4. No cross reactivity with other human heavy chains, mouse IgG, rat IgG, or goat IgG.
Target Species Human
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.