Learn More
Rabbit anti-Human IgG1 Fc Secondary Antibody (245), DyLight 350, Novus Biologicals™

Description
Specifications
Specifications
| Antigen | IgG1 Fc |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 245 |
| Conjugate | DyLight 350 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human IgG1 Fc (Uniprot#: P01857-1; Glu99-Lys330,C103S). HEK293 used as expression system. |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.