Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Human IgG3, Clone: RM119, Secondary Antibody,

Catalog No. 041654MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-165-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-165-4MI Supplier MilliporeSigma™ Supplier No. 041654
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifically detects Human IgG3 clone: RM119 in Human samples, and it is validated for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG3
Applications ELISA
Classification Monoclonal
Clone RM119
Formulation Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Host Species Rabbit
Immunogen Human IgG3
Purification Method Protein A Purfied
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary and Control Antibodies
Primary or Secondary Secondary
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt. Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.