Learn More
Sino Biological Rabbit anti-Human Kappa Light Chain Secondary Antibody, Rabbit MAb 100 µL
Catalog No. 5000786189
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant F(ab')2 of human kappa light chain IgG.
Catalog No. 50-007-86189 Supplier Sino Biological Supplier No. 68077R008100
This antibody was obtained from a rabbit immunized with purified, recombinant F(ab')2 of human kappa light chain IgG.