Learn More
Seracare Life Sciences Inc RABBIT ANTI-LEGIONELLA SPECIES
Supplier:  Seracare Life Sciences Inc 53300068
Encompass_Preferred
Protein G purified BacTrace antibody to Legionella species labeled with DyLight 488. Isolated from a serum pool of rabbits immunized with various strains of heat-killed Legionella.
Catalog No. 50-670-95