Learn More
ACROBiosystems Rabbit Anti-Mouse FMC63 scFv Polyclonal Antibody (recommended for ADA assay) (MALS verified)
Catalog No. 502054794
Encompass_Preferred
Antibody;100UG;Polyclonal;Recognizes FMC63 scFv specifically.;Rabbit Anti-Mouse FMC63 scFv Polyclonal Antibody (Cat. No. FM3-S93) is produced from a Rabbit immunized with FMC63 scFv.; ;Immobilized FMC63 scFv at 2 μg/mL (100 μL/well) can bind Rabbit Anti-Mouse FMC63 scFv Polyclonal Antibody (Cat. No. FM3-S93) with a linear range of 0.098-0.78 ng/mL (QC tested).;Immobilized Monoclonal Anti-FMC63 scFv Antibody at 1 μg/mL, add increasing concentrations of Rabbit Anti-Mouse FMC63 scFv Polyclonal Antibody and then add Biotinylated Monoclonal Anti-FMC63 scFv Antibody at 0.2 μg/mL. Detection was performed using HRP-conjugated streptavidin with sensitivity of 0.78 μg/mL (QC tested).
Catalog No. 50-205-4794 Supplier ACROBiosystems Supplier No. FM3S93100UG
Antibody;100UG;Polyclonal;Recognizes FMC63 scFv specifically.;Rabbit Anti-Mouse FMC63 scFv Polyclonal Antibody (Cat. No. FM3-S93) is produced from a Rabbit immunized with FMC63 scFv.; ;Immobilized FMC63 scFv at 2 μg/mL (100 μL/well) can bind Rabbit Anti-Mouse FMC63 scFv Polyclonal Antibody (Cat. No. FM3-S93) with a linear range of 0.098-0.78 ng/mL (QC tested).;Immobilized Monoclonal Anti-FMC63 scFv Antibody at 1 μg/mL, add increasing concentrations of Rabbit Anti-Mouse FMC63 scFv Polyclonal Antibody and then add Biotinylated Monoclonal Anti-FMC63 scFv Antibody at 0.2 μg/mL. Detection was performed using HRP-conjugated streptavidin with sensitivity of 0.78 μg/mL (QC tested).