Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit anti-Mouse IgG Fc Secondary Antibody, Biotin

Catalog No. PI31813 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31813 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31813 Supplier Invitrogen™ Supplier No. 31813
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31813 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31813 reacts with the heavy chains of mouse IgG, but not with the light chains common to most mouse immunoglobulins. This antibody does not react against mouse IgM, or against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Reconstitute with 1.5 mL of distilled water. Country of Origin: USA.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG Fc
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Biotin
Formulation PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Host Species Rabbit
Purification Method Affinity chromatography
Quantity 1.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.