Learn More
Rabbit anti-Mouse IgG gamma 1 heavy chain, HRP, Polyclonal, Secondary Antibody, Abnova™
Shop All Abnova Corporation ProductsSpecifications
Specifications
| Antigen | Rabbit anti-Mouse IgG gamma 1 heavy chain |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | HRP |
| Description | Rabbit anti-mouse IgG recognizes mouse IgG gamma 1 heavy chain. This secondary antibody was purified using antigen affinity chromatography. |
| Dilution | ELISA (1:300000) Western Blot (1:2000-1:10000) Immunohistochemistry (1:500-1:2500) The optimal working dilution should be determined by the end user. |
| Formulation | Lyophilized from 20mM potassium phosphate, 150mM NaCl, pH 7.2 (10mg/mL B SA, 0.01% (w/v) gentamicin sulfate). |
| Host Species | Rabbit |
| Immunogen | Mouse IgG gamma 1 heavy chain |
| Purification Method | This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.