Learn More
Immunology Consultants anti-Mouse IgG h+l Antibody (Rabbit) - Affinity Purified
SDP
Catalog No. 502248881
Encompass_Preferred
RGHL‑90A is an affinity‑purified rabbit antibody specific for mouse IgG heavy and light chains, designed for use as a secondary antibody in immunoassays. This reagent provides strong, selective binding and low background, supporting reliable detection in ELISA, Western blotting, and other immunochemical applications. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑90A is an essential tool for researchers working with murine antibodies.
This item is not returnable. View return policy
Catalog No. 50-224-8881 Supplier Immunology Consultants Supplier No. RGHL90A
RGHL‑90A is an affinity‑purified rabbit antibody specific for mouse IgG heavy and light chains, designed for use as a secondary antibody in immunoassays. This reagent provides strong, selective binding and low background, supporting reliable detection in ELISA, Western blotting, and other immunochemical applications. Produced by Immunology Consultants and supplied in a 1 mg format, RGHL‑90A is an essential tool for researchers working with murine antibodies.