Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, MP Biomedicals

Catalog No. ICN55480 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55480 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55480 Supplier MP Biomedicals, Inc Supplier No. 0855480
Add to Cart
Add to Cart

Host Species: Rabbit
Species Reactivity: Mouse

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG, Whole Molecule
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
Quantity 2 mg
Primary or Secondary Secondary
Target Species Mouse
Form Purified
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.