Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Rabbit anti-Mouse IgG1, Clone: RM106, Secondary Antibody,

Catalog No. 041644MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
04-164-4MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-164-4MI Supplier MilliporeSigma™ Supplier No. 041644
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Monoclonal Antibody

Specifically detects IgG1 Clone: RM106 in Mouse samples, and it is validated for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1
Applications ELISA
Classification Monoclonal
Clone RM106
Conjugate Unconjugated
Formulation Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.
Host Species Rabbit
Immunogen Mouse IgG
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Secondary, Control Antibodies
Primary or Secondary Secondary
Test Specificity This antibody reacts to the Fc region of mouse IgG1. No cross reactivity with mouse IgG2a, IgG3, IgM, IgA, IgE, human IgG, or rat IgG. It may slightly cross react to mouse IgG2b, and may also react to goat IgG. The Fc region of RM106 has been engineered to eliminate the binding of Fc receptor.
Target Species Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.