Learn More
Sino Biological Rabbit Anti-Mouse IgG1 Secondary Antibody (HRP), Rabbit MAb 20 µg
Catalog No. 5000750865
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IgG1 (rM IgG1; Catalog# 10690-MNAH; P01868-1; Val98-Lys324,C102S), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-007-50865 Supplier Sino Biological Supplier No. 10690R020H20
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IgG1 (rM IgG1; Catalog# 10690-MNAH; P01868-1; Val98-Lys324,C102S), and conjugated with horseradish-peroxidase (HRP).