Learn More
Sino Biological Rabbit Anti-Mouse IgG1 Secondary Antibody, Rabbit MAb 20 µL
Catalog No. 5000750078
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IgG1 (rM IgG1; Catalog# 10690-MNAH; P01868-1; Val98-Lys324,C102S).
Catalog No. 50-007-50078 Supplier Sino Biological Supplier No. 10690R02020
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IgG1 (rM IgG1; Catalog# 10690-MNAH; P01868-1; Val98-Lys324,C102S).