Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Mouse IgG2a Heavy Chain Secondary Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB483844
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB483844 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB483844 Supplier R&D Systems Supplier No. NBP172794UV

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG2a Heavy Chain Polyclonal specifically detects IgG2a Heavy Chain in Mouse samples. It is validated for Dot Blot, ELISA, Flow Cytometry, Fluorophore-linked immunosorbent assay, Immunohistochemistry, Western Blot.

Specifications

Antigen IgG2a Heavy Chain
Applications Dot Blot, ELISA, Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Host Species Rabbit
Immunogen Mouse IgG2a heavy chain
Purification Method Multi-step
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.