Learn More
Rabbit anti-Mouse IgG2a Heavy Chain Secondary Antibody, Texas Red, R&D Systems™

Description
Specifications
Specifications
| Antigen | IgG2a Heavy Chain |
| Applications | Flow Cytometry, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Texas Red |
| Dilution | Western Blot 1:1000 - 1:10000, Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 |
| Host Species | Rabbit |
| Immunogen | Mouse IgG2a heavy chain |
| Purification Method | Affinity Purified |
| Quantity | 1 mg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.