Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-Mouse IgG2b Heavy Chain Secondary Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP172892
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP172892 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP172892 Supplier R&D Systems Supplier No. NBP172892

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

IgG2b Heavy Chain Polyclonal specifically detects IgG2b Heavy Chain in Mouse samples. It is validated for Dot Blot, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen IgG2b Heavy Chain
Applications Dot Blot, Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate DyLight 488
Dilution Western Blot 1:10000, Immunocytochemistry/Immunofluorescence 1:5000, Dot Blot, Fluorophore-linked immunosorbent assay 1:20000
Host Species Rabbit
Immunogen Mouse IgG2b chain
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Secondary
Test Specificity Minimal cross reactivity was noted against other Mouse immunoglobulin classes or subclasses.
Reconstitution Reconstitute with 100 ul deionized water (or equivalent).
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.