Learn More
Rabbit anti-Mouse IgG3 Heavy Chain Secondary Antibody, DyLight 405, R&D Systems™

Description
Specifications
Specifications
| Antigen | IgG3 Heavy Chain |
| Applications | Fluorophore-Linked Immunosorbent Assay, Immunocytochemistry/Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | DyLight 405 |
| Dilution | Western Blot, Fluorophore-linked immunosorbent assay |
| Gene Alias | DKFZp686H11213, FLJ39988, FLJ40036, FLJ40253, FLJ40587, FLJ40789, FLJ40834, IgG3, immunoglobulin heavy constant gamma 3 (G3m marker), MGC45809 |
| Gene Symbols | IGHG3 |
| Host Species | Rabbit |
| Immunogen | Mouse IgG3 chain |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.