Learn More
Enzo Life Sciences Rabbit anti-Mouse Linker (Ready-to-use) (6ml)
SDP
Catalog No. 502013364
Encompass_Preferred
The Rabbit anti-Mouse Linker is a ready-to-use signal amplifying reagent optimized to increase the detection sensitivity of Enzo's POLYVIEW PLUS (anti-Rabbit) Reagents. Isotype: IgG. Species reactivity: Mouse . Applications: IHC ISH (in situ hybridization). Formulation: Ready-to-use solution in PBS containing stabilizers and preservatives.
Catalog No. 50-201-3364 Supplier Enzo Life Sciences Supplier No. ENZACC1166000
The Rabbit anti-Mouse Linker is a ready-to-use signal amplifying reagent optimized to increase the detection sensitivity of Enzo's POLYVIEW PLUS (anti-Rabbit) Reagents. Isotype: IgG. Species reactivity: Mouse . Applications: IHC ISH (in situ hybridization). Formulation: Ready-to-use solution in PBS containing stabilizers and preservatives.